Human HSP90 ELISA Kit (Chemiluminescence)
Lieferant: NOVUS BIOLOGICALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
NBP2-76643
NOVUNBP2-76643EA 1150 EUR InStock
NOVUNBP2-76643
Human HSP90 ELISA Kit (Chemiluminescence)
Tests ELISAs
This kit recognizes Human HSP-90 in samples. No significant cross-reactivity or interference between Human HSP-90 and analogues was observed.
  • Standard curve range: 62,5 to 4000 pg/ml
  • Inter-Assay: CV% <10,47%
  • Intra-Assay: CV% <9,68%
  • Spike Recovery: 91 to 117%
  • Gene: HSP90AA1
Hsp90 has at least 2 isoforms, Hsp90a and Hsp90b which are encoded by separate genes. These ubiquitous and highly conserved proteins account for 1 to 2% of all cellular proteins in most cells. Hsp90 is part of the cells powerful network of chaperones to fight the deleterious consequences of protein unfolding caused by nonphysiological conditions. However, in the absence of stress, Hsp90 is a necessary component of fundamental cellular processes such as hormone signaling and cell cycle control. In this context several key regulatory proteins such as steroid receptors, cell cycle kinases involved in signal transduction and p53 have been identified as substrates of Hsp90. It has been suggested that Hsp90 acts as a capacitor for morphological evolution by buffering widespread variation, which may affect morphogenic pathways.
Lieferumfang: Kit contains Biotinylated detection Ab diluent, Concentrated biotinylated detection Ab (100x), Concentrated HRP conjugate (100x), Concentrated wash buffer (25x), HRP conjugate diluent, Micro CLIA plate (dismountable), Plate sealer, Product manual, Reference standard, Sample diluent, Substrate reagent A and B.
Achtung! Bitte beachten: For research use only and is not approved for use in humans or in clinical diagnosis.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge