Closed top caps for storage vials
Lieferant: THERMO NATIONAL SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
B7815-13 B7815-24 B7815-20
NSCAB7815-13EA 67.6 EUR InStock
NSCAB7815-13 NSCAB7815-24 NSCAB7815-20
Closed top caps for storage vials
Verschlüsse Chromatographieampullen-Verschlüsse
PTFE on film/foam backing offers broad chemical resistance.
Not autoclavable.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge