Barcode reader, Nunc™
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
330050
NUNC330050EA 598 EUR InStock
NUNC330050
Barcode reader, Nunc™
Barcode- und RFID-Systeme
This durable barcode reader is ideal for simple track sampling. It is for use with any USB slot.
  • 'Plug and Play' function can automatically switch to read codes: 128A, 128B, 128C, 39 and interleave ²/₅ Convenient and easy to use with an Excel spread sheet
  • Scanner is a contact scanner which means it must be in contact with the barcode to read
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge