Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ölrot O 0,5% in 2-Propanol (IPA), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

O1391-250ML O1391-500ML
O1391-250MLEA 46.7 EUR InStock
O1391-250ML O1391-500ML
Ölrot O 0,5% in 2-Propanol (IPA), Sigma-Aldrich®
Ölrot O
Oil Red O stains lipid materials and imparts red-orange color to the lipids. It also marks vacuoles, particularly observed in ALL-L3 (acute lymphoblastic leukemia) or Burkitt′s lymphoma/leukemia. Oil Red O staining is done on fresh samples, as alcohol fixation removes lipids.

Oil red O solution has been used to stain neutral lipids and adipocytes and to visualize fat droplets in fibroblasts. Oil red O is a lysochrome (fat-soluble dye) diazo dye used for staining of neutral triglycerides and lipids on frozen sections and some lipoproteins on paraffin sections. In histology, a supersaturated solution of oil red O in isopropanol may be used to stain fat in tissue.


Formel: C₂₆H₂₄N₄O
MW: 408.51 g/mol
Flash-Pt: 12.0 °C (geschlossener Tiegel)
Storage Temperature: Room Temperature
MDL Number: MFCD00003898
CAS-Nummer: 1320-06-5
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12a6547340ed0b3a1466a0a2e6d2e16cc1a3f0c4&vid=12a6547340ed0b3a1466a0a2e6d2e16cc1a3f0c4&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance
dark red, sediment free
Conforms
Absorbance
diluted 0.25ml to 50 ml with Isopropyl alcohol
0.20 - 0.50
Lambda Max 510 - 517 nm

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge