Guardian 7000 magnetic hotplate stirrers
Lieferant: OHAUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30500603 30500613 30500623
OHAU30500603EA 1050 EUR InStock
OHAU30500603 OHAU30500613 OHAU30500623
Guardian 7000 magnetic hotplate stirrers
Rührer Magnetrührer mit Heizplatte
Durable magnetic hotplate stirrers with superior safety features and intelligent performance.
  • SmartPresence™ and SmartLink™ technologies allow unsupervised operation
  • SafetyHeat™ system protects the laboratory
  • Powerful, consistent stirring performance
  • Customised performance for precise applications
OHAUS Guardian 7000 magnetic hotplate stirrers are designed with outstanding safety features and superior heating and mixing performance. The SmartHousing™ is chemical resistant with an innovative, easy-to-clean design that channels spills away from internal components and the angled glass control panel.
The bright LCD display includes intuitive icons as well as temperature, speed and time settings – all designed to monitor performance from across the laboratory.
Lieferumfang: Supplied with temperature probe.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge