Bood DNA HV kit, CE IVD, Mag-Bind®
Lieferant: OMEGA BIOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M3292-03CEIVD
OMEGM3292-03CEIVDEA 2720 EUR InStock
OMEGM3292-03CEIVD
Bood DNA HV kit, CE IVD, Mag-Bind®
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
The Mag-Bind® Blood DNA HV Kit CE IVD is designed for rapid and reliable isolation of high-quality genomic DNA from up to 4 ml whole blood samples.
The Mag-Bind® Blood DNA HV Kit CE IVD is designed for rapid and reliable isolation of high-quality genomic DNA from up to 4 ml whole blood samples. Mag-Bind® Particles CH provide a quick magnetic response time reducing overall processing time. This system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with the time-proven efficiency of Omega Bio-Tek’s DNA isolation system to provide a fast and convenient method to isolate DNA. Utilizing paramagnetic particles provides high-quality DNA that is suitable for direct use in most downstream applications, such as amplification and enzymatic reactions. The 4 ml whole blood protocol has been successfully automated on the Hamilton Microlab® STAR™.
Zertifizierungen: CE-IVD IVDR
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge