Viral DNA/RNA Xpress kit, CE IVD, Mag-Bind®
Lieferant: OMEGA BIOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M62192304CEIVD
OMEGM62192304CEIVDEA 5210 EUR InStock
OMEGM62192304CEIVD
Viral DNA/RNA Xpress kit, CE IVD, Mag-Bind®
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Mag-Bind® Viral DNA/RNA Xpress Kit CE IVD follows a magnetic bead-based approach for the rapid and reliable isolation of viral DNA and RNA from nasopharyngeal (NP) swab specimens that are dry or in viral transport media (VTM), from saliva and other sample sources.
Mag-Bind® Viral DNA/RNA Xpress Kit CE IVD follows a magnetic bead-based approach for the rapid and reliable isolation of viral DNA and RNA from nasopharyngeal (NP) swab specimens that are dry or in viral transport media (VTM), from saliva and other sample sources. The extraction methodology is easily adaptable to various automated systems and can also be scaled up or down depending on the amount of starting sample amount used. The kit utilizes the proven Mag-Bind® technology that enables the purification of high-quality nucleic acids that are free of proteins, nucleases, and other impurities. The purified nucleic acids are ready for direct use in downstream applications such as qPCR, RT-qPCR, and more.
Zertifizierungen: CE-IVD IVDR
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge