Mag-Bind® FFPE DNA/RNA 96 Kit
Lieferant: OMEGA BIOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M6955-00 M6955-01
OMEGM6955-00EA 691 EUR InStock
OMEGM6955-00 OMEGM6955-01
Mag-Bind® FFPE DNA/RNA 96 Kit
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Sequentially isolate DNA and RNA in two separate eluates from the same FFPE sample.
  • Xylene-free protocol
  • Extraction of both DNA and RNA in separate eluates from the same FFPE sample
  • No splitting of samples
  • Magnetic bead-based purification
  • Automation friendly
Mag-Bind® FFPE DNA/RNA 96 Kit is designed for the sequential isolation of DNA and RNA from the same formalin-fixed, paraffin-embedded (FFPE) tissue sample. The protocol utilises non-toxic mineral oil in the place of the hazardous xylene for efficient deparaffinisation of the FFPE sample. The specially formulated buffers reverse cross-linking without the need for overnight digestion resulting in high-yielding, high-quality nucleic acids.
The isolation protocol allows for the extraction of both DNA and RNA in separate eluates from the same sample for a comprehensive analysis of both the nucleic acids. Purified DNA and RNA are suitable for a variety of downstream applications including SNP analysis, sequencing, and genotyping. The Mag-Bind® system is fully automatable on the Hamilton Microlab® STAR™, Tecan Fluent™, Thermo KingFisher® Flex™, and other open-ended workstations.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge