VHB DNA Washing Buffer
Lieferant: OMEGA BIOTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHB-440 VHB-1760
OMEGVHB-440EA 527 EUR InStock
OMEGVHB-440 OMEGVHB-1760
VHB DNA Washing Buffer
Puffer Elektrophorese-Puffer Waschpuffer
VHB buffer must be diluted with ethanol before use.
Achtung! Bitte beachten: For Research Use Only. Not for use in diagnostic procedures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge