Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Rotary valves, high pressure

Lieferant: OMNIFIT

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

001120 001121 001122 001126
OMNI001120EA 982 EUR InStock
OMNI001120 OMNI001121 OMNI001122 OMNI001126
Rotary valves, high pressure
Ventile Schaltventile
The high pressure manual rotary valves are made from PTFE with a PCTFE rotor with a bore size of 0,8 mm making them ideal for pressure applications up to 500 psi. They are available in four and five port versions. All ports are 1/4"-28 UNF flat-bottom and will accept any 1/4"-28 UNF male fitting. These valves are mountable using the pre-drilled M3 mounting holes.

  • Pressure rated up to 500 psi
  • Accept 1/4"-28 UNF fittings
  • M3 mounting holes

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge