Watertight Hard Case, CS-01
Lieferant: OPTIKA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CS-01
OPTKCS-01EA 224 EUR InStock
OPTKCS-01
Watertight Hard Case, CS-01
Beutel Equipment Bags and Instrument Cases
CS-01 is a watertight hard case, which is made to offer superior protection from the elements, water, dust and impacts when transporting fragile and valuable objects, like a microscope.
Convoluted foam in the lid and cubed foam (three layers) in the bottom with pick and pluck system to easely adapt the case to different microscopes.
It can be used for carrying following models:
– B-190 Series: B-191PL, B-192PL, B-193PL
– B-290 Series: B-292, B-292PLI, B-293, B-293PLI
– B-380 Series: B-382. B-382PLI, B-383, B-383PLI, B-382PH, B-382PHI, B-383PH, B-383PHI, B-383DK
– B-510 Series: B-510BF, B-510Ergo, B-510PH, B-510DK
– SLX Series: SLX-1, SLX-2, SLX-3
– SZ Series: all models equipped with bases SZ-ST1, SZ-ST2, SZ-ST3, SZ-ST7, SZ-ST8
Zertifizierungen: IP 67, ATA 300 and military certified.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge