Laboratory Microscope, SZR-180
Lieferant: OPTIKA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SZR-180
OPTKSZR-180EA 10900 EUR InStock
OPTKSZR-180
Laboratory Microscope, SZR-180
Mikroskope
The SZR-180 is the top-of-the-range stereo zoom microscope dedicated to the world of research.
  • Dioptric adjustment: Both eyepieces
  • Objective: PLAN Apochromatic 1×, N.A. 0,15, 7,5-135× (zoom ratio 18:1), zoom click stops at 0,75, 1, 2, 3, 6, 10, 13,5× with aperture diaphragm
  • Head: Trinocular 100/0 to 0/100 (on right tube), 20° inclined
  • Focusing: Coaxial coarse & fine, coarse total travel 125 mm, fine single rotation 2 mm, rack and pinion focusing mechanism
With a wide magnification range and high optical resolution combined with effective ergonomics, the SZR-180 is the perfect companion in advanced research. The 18:1 zoom allows you to observe a wide range of samples, from single cells to entire microorganisms.
The SZR-180 with its 18:1 zoom ratio puts it at the top of its class. The wide range of magnifications available (7,5 - 135×), coupled with a precise click-stop mechanism for working with reproducible zoom settings, makes it ideal for both low-magnification observation and high-magnification screening and observation of small cell structures.
The Plan APO 1× apochromatic main objective provides high-resolution images at both low and high magnifications, making it ideal for observing and capturing images of the smallest details free of aberrations and halos. Combined with the zoom and the generous 23 mm field number eyepieces, this provides a large FOV even at high magnifications (1,7 mm).
Ergonomically designed, the SZR-180’s wide base offers a huge space for sample holders, intuitive controls and an extremely low profile for easy access and reduced fatigue-free body movement. The LED diascopic illumination is designed to achieve high overall contrast and includes the innovative OIC (Oblique Illumination Contrast) system, ideal for transparent structures, and dark-field illumination.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge