pH electrode, double junction, Orion™ 9110DJWP
Lieferant: Thermo Orion
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9110DJWP
ORIO9110DJWPEA 266 EUR InStock
ORIO9110DJWP
pH electrode, double junction, Orion™ 9110DJWP
Elektroden pH-Elektroden
This double junction pH electrode with semi-micro tip measures samples as small as 0,2 ml.
  • With waterproof BNC connector
  • Ag/AgCl double junction
  • Refillable
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge