Ion selective electrode, half-cell, fluoroborate, Orion™
Lieferant: Thermo Orion
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9305BN
ORIO9305BNEA 1730 EUR InStock
ORIO9305BN
Ion selective electrode, half-cell, fluoroborate, Orion™
Elektroden ISE-Elektroden
This fluoroborate half-cell can be easily and economically rejuvenated.
Plastic membrane half-cell requires reference electrode.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge