Silver billet electrode, Orion™
Lieferant: Thermo Orion
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9780SC
ORIO9780SCEA 287 EUR InStock
ORIO9780SC
Silver billet electrode, Orion™
Elektroden Elektroden, sonstige
These electrodes are a combination of a silver billet-indicating electrode along with a silver billet half-cell. Ideal for obtaining an accurate determination of potentiometric halides with dual electrodes.
Internal reference: Ag/AgCl
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge