Test kits in ampoules, AQUAfast™ IV reagents, Orion™
Lieferant: THERMO ORION
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AC4004 AC4005 AC4006 AC4007 AC4011 AC4012 AC4016 AC4017 AC4046 AC4048 AC4055 AC4060 AC4065 AC4070 AC4078 AC4082 AC4095 AC4099
ORIOAC4004EA 211 EUR InStock
ORIOAC4004 ORIOAC4005 ORIOAC4006 705-0751 ORIOAC4011 ORIOAC4012 ORIOAC4016 ORIOAC4017 ORIOAC4046 ORIOAC4048 ORIOAC4055 ORIOAC4060 ORIOAC4065 705-0752 ORIOAC4078 ORIOAC4082 705-0753 ORIOAC4099
Test kits in ampoules, AQUAfast™ IV reagents, Orion™
Umwelt-Prüfkits
Orion™ AQUAfast™ Auto-Test Ampoule Reagents can be used with the Orion™ AQUAfast™ AQ4000 colorimeter to simplify sample preparation and tests.
  • Consistency and reproducibility improved with AQUAfast™ ampoules and an AQ4000 meter
  • Auto-ID™ eliminates measurement errors
  • Designed for use with colorimetric water and wastewater testing equipment
The Auto-Test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge