Test kits for water testing, AQUAfast™ Powder Packs, Orion™
Lieferant: THERMO ORION
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AC4P12 AC4P29 AC4P42 AC4P46 AC4P55 AC4P60 AC4P71 AC4P72 AC4P78 AC4P79 AC4P82 AC4P95 AC4P54
ORIOAC4P12EA 411 EUR InStock
ORIOAC4P12 ORIOAC4P29 ORIOAC4P42 ORIOAC4P46 ORIOAC4P55 ORIOAC4P60 ORIOAC4P71 ORIOAC4P72 ORIOAC4P78 ORIOAC4P79 ORIOAC4P82 ORIOAC4P95 ORIOAC4P54
Test kits for water testing, AQUAfast™ Powder Packs, Orion™
Umwelt-Prüfkits
Orion™ AQUAfast Powder Pack reagents perform colorimetric measurements of common water parameters with powders that dissolve quickly and mix easily.
  • Equivalent to other widely used and recognized chemistries
  • Compatible with Orion™ AQUAfast Colorimeters
Orion™ AQUAfast Powder Pack Reagents are designed for use with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Powder pack chemistries dissolve quickly in solution, are easy to mix, and do not require a stir rod or crushing. Each powder pack is notched for simplified opening in the lab or field.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge