AC4P12
AC4P29
AC4P42
AC4P46
AC4P55
AC4P60
AC4P71
AC4P72
AC4P78
AC4P79
AC4P82
AC4P95
AC4P54
ORIOAC4P12EA
411
EUR
InStock
ORIOAC4P12
ORIOAC4P29
ORIOAC4P42
ORIOAC4P46
ORIOAC4P55
ORIOAC4P60
ORIOAC4P71
ORIOAC4P72
ORIOAC4P78
ORIOAC4P79
ORIOAC4P82
ORIOAC4P95
ORIOAC4P54
Test kits for water testing, AQUAfast™ Powder Packs, Orion™
Umwelt-Prüfkits
Orion™ AQUAfast Powder Pack reagents perform colorimetric measurements of common water parameters with powders that dissolve quickly and mix easily.
- Equivalent to other widely used and recognized chemistries
- Compatible with Orion™ AQUAfast Colorimeters
Orion™ AQUAfast Powder Pack Reagents are designed for use with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Powder pack chemistries dissolve quickly in solution, are easy to mix, and do not require a stir rod or crushing. Each powder pack is notched for simplified opening in the lab or field.