Hard-sided field case for portable meters, Orion™ Star A series
Lieferant: Thermo Orion
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
STARA-CS
ORIOSTARA-CSEA 144 EUR InStock
ORIOSTARA-CS
Hard-sided field case for portable meters, Orion™ Star A series
Elektrodenhalter
This hard-sided field case can be used to protect, transport and store measuring equipment in the field.
The field case holds Star A series portable meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case plus the internal separating door can be removed for cable management.
For use with: Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge