pH measurement module for Orion™ Versa Star Pro™ meters
Lieferant: Thermo Orion
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VSTAR-PH
ORIOVSTAR-PHEA 542 EUR InStock
ORIOVSTAR-PH
pH measurement module for Orion™ Versa Star Pro™ meters
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This measurement module can be used to customise the measurement capabilities of the Orion™ Versa Star Pro™ meters. It adds pH, mV, ORP and temperature measurement capabilities to the meter.
  • Each Orion™ Versa Star Pro meter has four channels that can be customised with interchangeable measurement modules
  • Measurement modules are interchangeable
  • Up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customise measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
Range (pH): −2,000 to +20,000
Range (mV): ±2000,0
Range (Temperature): −5,0 to +105,0 °C
3-year replacement warranty.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge