Filling System, RF-3
Lieferant: THERMO OWL SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RF-3
OWLARF-3EA 406 EUR InStock
OWLARF-3
Filling System, RF-3
Möbel Regale
Organize your refrigerator, coldroom, freezer or benchtop with the Thermo Scientific™ Owl™ RF-3 Filing System.
  • Eight compartments constructed of durable 640 mm thick acrylic
  • No-skid rubber feet prevent sliding
  • Use either vertically or horizontally
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge