Oxoid™ AnaeroBox™ Box with Lid and Seal
Lieferant: THERMO FISHER DIAGNOSTICS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AB0035A
OXOIAB0035AEA 193 EUR InStock
OXOIAB0035A
Oxoid™ AnaeroBox™ Box with Lid and Seal
Anaerobe Systeme
Culture anaerobic and microaerophilic organisms by achieving the right atmospheric conditions using Oxoid™ AnaeroBox™ 3,5 L box with lid and seal.
The Box is designed to be used with Oxoid™ AnaeroGen™ 3,5 L sachet and Oxoid™ CampyGen™ 3,5 L sachet. The AnaeroGen 3,5 L Sachet produces an anaerobic atmosphere rapidly without the use of water or catalyst. No hydrogen is produced. The oxygen is absorbed rapidly without the build-up of pressure. The final atmosphere contains less than 1% oxygen supplemented with carbon dioxide - ideal for optimum growth of anaerobes and increased colony size. The CampyGen 3,5 L sachet produces a microaerophilic atmosphere rapidly without the use of water or catalyst. No hydrogen is produced. The oxygen is absorbed rapidly without the build-up of pressure. The final atmosphere contains an ideal microaerobic atmosphere of 8 to 9% oxygen and 7 to 8% carbon dioxide.
Bestellinformation: Oxoid™ AnaeroGen™ 3,5 L sachet and Oxoid™ CampyGen™ 3,5 L sachet are not included.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge