Anaerobic Incubation System, Anaerogen Jars
Lieferant: THERMO FISHER DIAGNOSTICS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AG0025A CN0025A CN0035A CN0020C AN0020D
OXOIAG0025AEA 361 EUR InStock
OXOIAG0025A OXOICN0025A OXOICN0035A OXOICN0020C OXOIAN0020D
Anaerobic Incubation System, Anaerogen Jars
Anaerobe Systeme
The jar is designed for use with AnaeroGen/CampyGen sachet and will hold up to 12 plates.
  • Both jar and lid are made of robust materials to ensure protection of operator and equipment
  • No catalyst required
  • A carrying handle for the safe transportation of the jar from bench to incubator
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge