Alum adjuvant, AlumVax hydroxide
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AH0050 AH0250
OZBIAH0050EA 119.9 EUR InStock
OZBIAH0050 OZBIAH0250
Alum adjuvant, AlumVax hydroxide
Protein- Expression Proteinexpressions-Reagenzien
AlumVax hydroxide is a wet gel (colloidal) of aluminium hydroxide 2%. It is the most common adjuvant used in approved prophylactic vaccines because of its excellent safety profile and ability to enhance the protective humoral immune response.
  • Ready to use suspension
  • Store at room temperature
It has been observed that aluminium compounds act by a depot effect and also by direct activation of the immune cells. Adsorption or entrapment of antigens in aggregates through hydrophobic and electrostatic interactions favours a high local antigen concentration and improved uptake by antigen presentating cells (APC).
AlumVax hydroxide is a crystalline aluminium oxyhydroxide that is positively charged at physiological pH (pI=11), suitable for adsorption of negatively charged, acidic proteins (such as albumin). It stimulates Th2 response through the release of Th2-associated cytokines (IL4, IL-5, IL-13) and Th2-associated antibodies (IgG1 and IgE). It increases Ag-specific CD4+ T Cell proliferation and promotes NALP3 inflammasome activation and caspase 1-mediated release of IL-1 and IL-18.
Lieferumfang: Shipped at room temperature.
Achtung! Bitte beachten: Intended for research use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge