Bradford protein assay kits (B-Pak)
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BA00050 BA00070 BA00100
OZBIBA00050EA 133.3 EUR InStock
OZBIBA00050 OZBIBA00070 OZBIBA00100
Bradford protein assay kits (B-Pak)
Tests Proteintests
Bradford Protein Assay Kit (B-Pak) is a straightforward and rapid kit for determining the concentration of proteins in solution. This ready to use kit is provided with 1X reagent and 2 sets of BSA standards which means no dilution, filtration or calculation required. B-Pak is provided with adapted protocols for all experimental conditions – micro and macroassays.
  • Accurate determination of protein concentration
  • Ready to use prediluted standard protein
  • Cheaper than competitors
  • Convenient packaging
  • Fast and easy to use
Store at +4 °C.
Lieferumfang: Shipped at room temperature.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge