Protein transfection reagent, Pro-DeliverIN CRISPR
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PIC60100 PIC60500 CAS9PIC
OZBIPIC60100EA 204 EUR InStock
OZBIPIC60100 OZBIPIC60500 OZBICAS9PIC
Protein transfection reagent, Pro-DeliverIN CRISPR
Transfektionsreagenzien
Pro-DeliverIN CRISPR is a transfection reagent optimised for recombinant Cas9 protein delivery or Cas9/gRNA RNP complexes. Suitable for gene editing applications, this reagent provides high transfection efficiency with minimal toxicity.
  • High efficiency delivery of Cas9 protein or Cas9 gRNA complexes
  • Low cell toxicity - enable to start experiment with less cells
The Cas 9 recombinant protein is delivered more rapidly than nucleic acid and is fully active once inside the cells without latency period (in contrast to transcription and translation machineries required for the nucleic acids). As for mRNA, it also offers the advantage of avoiding genomic integration and is cell cycle independent.
Efficient delivery represents a critical step for genome editing experiments.
Lieferumfang: Supplied with 100 µl of R-Phycoerythrin positive control.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge