DNA transfection reagent, DreamFect™
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DF40500 DF41000 DF45000
OZBIDF40500EA 171 EUR InStock
OZBIDF40500 OZBIDF41000 OZBIDF45000
DNA transfection reagent, DreamFect™
Transfektionsreagenzien
DreamFect™ transfection reagent is based on an innovative biochemical method. It contains both a lipidic part and a cationic part allowing the association of nucleic acids with this transfection reagent and their delivery inside cells. Due to its unique properties, it releases large amounts of DNA in the cytosol increasing DNA nuclear delivery (Tee-technology). Consequently, it is highly efficient at low doses of nucleic acids and less cationic lipids are used so potential secondary effects are avoided.
  • Exceptional efficiency
  • High reproducibility
  • Broad range of cell types
  • Non-toxic
  • Serum compatible
  • Simple, rapid, and easy to use
Store at +4 °C.
Lieferumfang: Shipped at room temperature.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge