DNA transfection reagent, EcoTransfect™
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ET10500 ET11000 ET13000
OZBIET10500EA 111.8 EUR InStock
OZBIET10500 OZBIET11000 OZBIET13000
DNA transfection reagent, EcoTransfect™
Transfektionsreagenzien
EcoTransfect™ transfection reagent is an economical lipofection reagent dedicated to the transfection of popular cell lines. It is the perfect solution for quickly analysing the biological activity of nucleic acids, to perform routine transfection assays at low cost and to accomplish high throughput screening.
  • Economical
  • Good efficiency
  • Serum compatible
  • Simple, ready to use and rapid
Store at +4 °C.
Lieferumfang: Shipped at room temperature.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge