Transfection reagents, FluoMag™
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FN10100 FC10100 FP10100 FS10100 FV10100
OZBIFN10100EA 134.38 EUR InStock
OZBIFN10100 OZBIFC10100 OZBIFP10100 OZBIFS10100 OZBIFV10100
Transfection reagents, FluoMag™
Transfektionsreagenzien
FluoMag™ reagents are fluorescently-labeled magnetic nanoparticles for Magnetofection™ applications. FluoMag™ transfection reagents are as efficient as their non labelled counterparts and allow visualisation of the nanoparticles in vitro during experiments.
  • Advantages of Magnetofection™ technology
  • Easy cell labelling and tracking
  • Non toxic
  • Serum compatible
Store at +4 °C and protect from the light.
Bestellinformation: This reagent needs to be used with a magnetic plate, available separately.
Lieferumfang: Shipped at room temperature.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge