ONPG assay kit
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
GO10001
OZBIGO10001EA 239.63 EUR InStock
OZBIGO10001
ONPG assay kit
Tests Enzymtests
ONPG assay kit is ideal to quantify high expression level of beta-Gal.
  • Convenient for all transfection assays
  • Suitable for cultured cells and tissues
  • Simple, rapid and easy-to-use
  • Economical
  • Optimised with stable buffers
All the necessary reagents are provided in this assay kit and offer a rapid, simple and sensitive method to quantify spectrophotometrically the enzyme expression level in LacZ transfected cells.
The levels of active β-Gal expression can be quickly measured by its catalytic hydrolysis of ONPG (o-nitrophenyl-beta-D-galactopyranoside) substrate to a bright yellow product (absorbance at 405 to 420 nm). LacZ is one of the most frequently reporter gene used in transfection experiments. The LacZ encoded protein (beta-galactosidase, beta-Gal), is very stable, resistant to proteolytic degradation and easily assayed.
Lieferumfang: Kit contains 5X lysis buffer (55 ml), standard dillution buffer (55 ml), ONPG substrate solution (55 ml), stop buffer (55 ml) and β-Gal enzyme standard (100 µl).
Achtung! Bitte beachten: Store all reagents at +4 °C except beta Gal enzyme.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..