Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

DNA transfection reagent, HYPE-5™

Lieferant: OZ BIOSCIENCES

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


HY01500 HY03000 HY15000 HY30000 HYR10003 HYR10015 HYR20030
OZBIHY01500EA 222.59 EUR InStock
OZBIHY01500 OZBIHY03000 OZBIHY15000 OZBIHY30000 OZBIHYR10003 OZBIHYR10015 OZBIHYR20030
DNA transfection reagent, HYPE-5™
Transfektionsreagenzien
HYPE-5™ transfection kit is dedicated to achieve high yield protein expression in mammalian cells. The kit has been designed for maximum recombinant protein expression in HEK293 and CHO cells grown in suspension. It is composed of the HYPE-5™ transfection reagent (a lipofection reagent) and HYPE-5™ Blast reagent (improves protein production in CHO cells). The HYPE-5™ transfection kit is ideal for bioreactor, spinner or flasks.

  • High yield recombinant protein production
  • Suitable for both HEK293 and CHO cells growing in suspension
  • Compatible with any synthetic or regular media used for protein production
  • Completely animal origin free

Store HYPE-5™ reagent at –20 °C and HYPE-5™ Blast at +4 °C.

Lieferumfang: Shipped at room temperature.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge