Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Freund's incomplete adjuvant, IFAVax

Lieferant: OZ BIOSCIENCES

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


IFA0010 IFA0050 IFA0100
OZBIIFA0010EA 47.59 EUR InStock
OZBIIFA0010 OZBIIFA0050 OZBIIFA0100
Freund's incomplete adjuvant, IFAVax
Protein- Expression Proteinexpressions-Reagenzien
Incomplete Freund’s Adjuvant (IFA) is a water-in-oil emulsion that is similar to the complete Freund’s Adjuvant (CFA) without addition of heat-killed mycobacteria (Mycobacterum butyricum).

  • Suitable for the stimulation of Th2 response and antibody production

It consists of a mixture of mineral oil and emulsifier in a ratio of 85% v/v oil and 15% v/v emulsifier. It is not a pre-formed emulsion and thus it must be mixed with an equal volume of aqueous solution of antigen and subsequently emulsified prior to use. The emulsifier is mannide monooleate, an ester consisting of mannitol as the hydrophilic residue and oleic acid (unsaturated C18 fatty acid) as the hydrophobic part.

Store at +4 °C

Lieferumfang: Shipped at room temperature.

Achtung! Bitte beachten: Intended for research use only.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge