Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Transfection reagent, in vivo DogtorMag™

Lieferant: OZ BIOSCIENCES

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


IV-DM30500 IV-DM31000 IV-TK30220 IV-KC30220
OZBIIV-DM30500EA 351.75 EUR InStock
OZBIIV-DM30500 OZBIIV-DM31000 OZBIIV-TK30220 OZBIIV-KC30220
Transfection reagent, in vivo DogtorMag™
Transfektionsreagenzien
In vivo DogtorMag™ transfection reagent has been designed for in vivo targeted transfection of various types of nucleic acids, such as DNA, RNA, oligonucleotides, using magnetic nanoparticles coated with synthetic lipids. DNA complexes can be easily administrated through various injection routes, such as systemic administration (intravenous, intra-artery) or local administration (intratumoral, intracerebroventricular).

  • Increased transfection efficiency
  • Targeted process (magnetically-driven)
  • Reduction of the systemic dissemination of vectors during injection
  • Reduction of the vector doses
  • Work under non permissive conditions (hypothermia, physiological flow conditions)
  • Penetration of the vector into tissues
  • Minimized toxicity
  • Universal – suitable for all nucleic acids

In vivo DogtorMag™ kit contains in vivo Dogtor transfection reagent plus in vivo CombiMag transfection reagent.

Store in vivo DogtorMag™ at –20 °C and in vivo CombiMag at +4 °C

Bestellinformation: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.

Lieferumfang: Shipped at room temperature.


Order Now

 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge