Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

siRNA transfection reagent, Lullaby™

Lieferant: OZ BIOSCIENCES

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


LL70500 LL71000 LL73000
OZBILL70500EA 210.87 EUR InStock
OZBILL70500 OZBILL71000 OZBILL73000
siRNA transfection reagent, Lullaby™
Transfektionsreagenzien
Lullaby™ is an ideal siRNA transfection reagent for gene silencing. Based on the TEE-technology, it has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and very low toxicity. RNA interference is a powerful technique to shut down genes expression in cells and organisms. This silencing effect constitutes a very helpful tool to study gene function and a promising approach for new therapeutic treatments.

  • Effective with very low doses of siRNA
  • Minimised off-target effects
  • Suitable for siRNA, shRNA, dsRNA and more
  • Applicable to a broad range of cells
  • Serum compatible and non-toxic
  • Straightforward protocol

Lullaby™ transfection reagent introduces siRNA duplexes in a variety of cells with a very high efficiency leading to exceptional knockdown effects with low doses of siRNA.

Store at +4 °C.

Lieferumfang: Shipped at room temperature.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge