LL70500
LL71000
LL73000
OZBILL70500EA
210.87
EUR
InStock
OZBILL70500
OZBILL71000
OZBILL73000
siRNA transfection reagent, Lullaby™
Transfektionsreagenzien
Lullaby™ is an ideal siRNA transfection reagent for gene silencing. Based on the TEE-technology, it has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and very low toxicity. RNA interference is a powerful technique to shut down genes expression in cells and organisms. This silencing effect constitutes a very helpful tool to study gene function and a promising approach for new therapeutic treatments.
- Effective with very low doses of siRNA
- Minimised off-target effects
- Suitable for siRNA, shRNA, dsRNA and more
- Applicable to a broad range of cells
- Serum compatible and non-toxic
- Straightforward protocol
Lullaby™ transfection reagent introduces siRNA duplexes in a variety of cells with a very high efficiency leading to exceptional knockdown effects with low doses of siRNA.
Store at +4 °C.
Lieferumfang: Shipped at room temperature.