Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

DNA transfection reagent, Magnetofectamine™ O2

Lieferant: OZ BIOSCIENCES

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


MTX2-0750 MTX2-1000
OZBIMTX2-0750EA 389.55 EUR InStock
OZBIMTX2-0750 OZBIMTX2-1000
DNA transfection reagent, Magnetofectamine™ O2
Transfektionsreagenzien
Magnetofectamine™ O2 is a powerful transfection kit designed for primary and hard-to-transfect cells. The alliance of MTX transfection reagent and CombiMag leads to increased transfection efficiency, minimised toxicity and enhanced gene expression. CombiMag, a magnetic formulation based on the Magnetofection™ technology, binds to MTX transfection reagent/DNA complexes and under the application of a magnetic field concentrates the genetic material onto cells and promotes cellular uptake. In this way, transfection efficiency is enhanced.

  • Less nucleic acids used - minimised toxicity
  • No need to change your standard protocol
  • Ideal for hard-to-transfect and primary cells
  • Simple, ready to use and rapid
  • Serum compatible and applicable to all nucleic acids

Storage : Upon reception and for long-term use, store the MTX REAGENT and MTXBoost at –20°C and CombiMag at +4°C.

Zubehörinformationen: This reagent needs to be used with a magnetic plate.

Lieferumfang: MTX2-1000 Starter Kit includes 1× super magnetic plate (MF1000) + 250 µl CombiMag + 750 µl MTX reagent + 3 ml MTX Boost 100X.
MTX2-0750 kit contains 250 µl CombiMag + 750 µl MTX reagent + 3 ml MTX Boost 100X (NB. does not include magnetic plate).


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge