DNA transfection reagent, PolyMag Neo™
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PG60100 PG60200 PG61000
OZBIPG60100EA 138.68 EUR InStock
OZBIPG60100 OZBIPG60200 OZBIPG61000
DNA transfection reagent, PolyMag Neo™
Transfektionsreagenzien
PolyMag Neo™ transfection reagent is specifically designed to achieve high transfection efficiency combined with superior transgene expression level. It is composed of magnetic nanoparticles coated with cationic molecules. It is the ideal transfection reagent for a wide variety of cells.
  • High transgene expression
  • High transfection efficiency
  • Multipurpose (various types of nucleic acids)
  • Universal (primary cells and cell lines)
  • Simple, ready to use and rapid
  • Non toxic
  • Compatible with and without serum-containing culture media
Store at +4 °C.
Zubehörinformationen: This reagent needs to be used with a magnetic plate.
Lieferumfang: Shipped at room temperature.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge