DNA transfection reagent, PolyMag™
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PN30200 PN31000 PN30100
OZBIPN30200EA 241.76 EUR InStock
OZBIPN30200 OZBIPN31000 OZBIPN30100
DNA transfection reagent, PolyMag™
Transfektionsreagenzien
PolyMag™ transfection reagent, based on Magnetofection™ transfection technology, has been optimised for use with all types of nucleic acids. The reagent is composed of magnetic nanoparticles coated with cationic molecules. It has been successfully tested on a large range of primary cells, hard to transfect cells and cell lines and its efficiency has been reported in many publications.
  • Highly efficient for primary cells
  • Powerful for hard to transfect cells
  • Non toxic
  • Serum-compatible
  • Simple, rapid and easy to use
  • For all types of nucleic acids
Store at +4°C.
Zubehörinformationen: This reagent needs to be used with a magnetic plate.
Lieferumfang: Shipped at room temperature.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge