SEAP assay kit
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SP00500
OZBISP00500EA 152.65 EUR InStock
OZBISP00500
SEAP assay kit
Tests Zelltests
SEAP assay kit (Secreted Alkaline Phosphatase) is a colorimetric assay for sensitive quantification of SEAP in culture medium from transfected cells or tissues.
  • Designed to detect SEAP reporter gene expression
  • Fast and easy analysis
  • Colorimetric-measure with a standard spectrophotometer or an ELISA reader (405 nm)
  • Multiple samples can be analysed in low volumes
  • Convenient and economical packaging: no tablet, no powder
Alkaline phosphatase cleaves the colourless 4-nitrophenylphosphate (pNPP) substrate to give a yellow p-nitrophenolate product in alkaline solutions. It is commonly used as a reporter gene expression and can be accurately quantified. It is a truncated form of placental alkaline phosphatase (GPI-anchored protein).
Lieferumfang: Kit contains pNPP-containing assay buffer component A (10×1 ml), reaction buffer component B (125 ml), reaction buffer component C (125 µl) and dilution buffer component D (50 ml).
Achtung! Bitte beachten: Store at +4 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge