Stem transfection reagents, DreamFect™ Stem
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ST30500 ST31000
OZBIST30500EA 232.17 EUR InStock
OZBIST30500 OZBIST31000
Stem transfection reagents, DreamFect™ Stem
Transfektionsreagenzien
DreamFect™ Stem is a powerful transfection reagent allowing multipotent stem cell transfection with high efficiency and very low toxicity. Its specific composition based on TEE-technology allows transfecting embryonic and multipotent stem cells in the presence of serum, maintaining their undifferentiated stage and their capacity to differentiate.
  • High transfection efficiency for multipotent stem cells
  • Minimised toxicity due to reagent biodegradability and low DNA amount required
  • Cell phenotype and differentiation potential are not affected
  • Serum compatible
  • Simple, ready to use and rapid
Store at –20 °C.
Lieferumfang: Shipped at room temperature.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge