TF20250
TF20500
TF21000
OZBITF20250EA
121.48
EUR
InStock
OZBITF20250
OZBITF20500
OZBITF21000
DNA transfection reagent, 3D-Fect™
Transfektionsreagenzien
3D-Fect™ transfection reagent is specifically designed to directly transfect cells cultured in 3D scaffolds.
- Highly efficient for cell lines and primary cells
- Specific for 3D scaffolds
- Compatible with all types of nucleic acids
- Long term protein expression
- Serum compatible
Scaffold-based 3D matrices activated with 3D-Fect™/DNA complexes are colonised by cells which are subsequently transfected in situ and in a more natural environment. This method allows studying tissue engineering and regeneration, tumour invasion, neural differentiation, cellular polarisation, tissue formation, colonisation, neurite growth, etc.
This method allows studying tissue engineering and regeneration, tumour invasion, neural differentiation, cellular polarisation, tissue formation,colonisation, neurite growth.