DNA transfection reagent, 3D-Fect™
Lieferant: OZ BIOSCIENCES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TF20250 TF20500 TF21000
OZBITF20250EA 121.48 EUR InStock
OZBITF20250 OZBITF20500 OZBITF21000
DNA transfection reagent, 3D-Fect™
Transfektionsreagenzien
3D-Fect™ transfection reagent is specifically designed to directly transfect cells cultured in 3D scaffolds.
  • Highly efficient for cell lines and primary cells
  • Specific for 3D scaffolds
  • Compatible with all types of nucleic acids
  • Long term protein expression
  • Serum compatible
Scaffold-based 3D matrices activated with 3D-Fect™/DNA complexes are colonised by cells which are subsequently transfected in situ and in a more natural environment. This method allows studying tissue engineering and regeneration, tumour invasion, neural differentiation, cellular polarisation, tissue formation, colonisation, neurite growth, etc.
This method allows studying tissue engineering and regeneration, tumour invasion, neural differentiation, cellular polarisation, tissue formation,colonisation, neurite growth.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge