Embedding medium, PolyFreeze, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P0091-6X120ML
P0091-6X120MLEA 319 EUR InStock
P0091-6X120ML
Embedding medium, PolyFreeze, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
PolyFreeze is a water-soluble support matrix and a form of embedding medium for frozen sectioning. PolyFreeze sections flawlessly at routine frozen specimen thickness of 5-6 μ and leaves no slide residue. It provides for -20 ºC cryostat sectioning in rapid set embedment. This medium freezes quickly supporting the tissue for sectioning at 3 mm and up with no cracking of the matrix at temperatures from -8 °C to -25 °C.
  • Embedding medium for frozen sectioning of plant and animal samples.
  • Sections flawlessly at routine frozen specimen thickness of 5 - 6 μ
  • Leaves no slide residue
  • Provides for –20 ºC cryostat sectioning in rapid set embedment
Snap freeze using PolyFreeze with isopentane and liquid nitrogen, or dry ice (slush/slurry or bunker). Store specimens frozen in PolyFreeze in liquid nitrogen canisters or in airtight containers in a -80 °C freezer. PolyFreeze does not cause autofluorescence and can easily be washed away during fixation and rinsed prior to staining. Once rinsed, there is no trace of the support matrix to interfere with staining or immunohistochemistry (IHC) reactions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge