Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Proflavine hemisulphate hydrate, Pulver, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ100

P2508-10G P2508-25G
P2508-10GEA 61.1 EUR InStock
P2508-10G P2508-25G
Proflavine hemisulphate hydrate, Pulver, Sigma-Aldrich®
Proflavine hemisulphate hydrate
Proflavine is a small, amphipathic, acridine-derived fluorescent dye.Proflavine hemisulfate is used as a rapid stain for cytological studies of biological specimens. It stains cytoplasmic structures and nuclei. Proflavine has the capability to intercalate double stranded DNA.

Proflavine hemisulfate salt hydrate has been used to stain cells of the 1483 cell line. It has also been used as a calibrating dye in fluorescence resonance energy transfer (FRET) analysis.


Formel: (C₁₃H₁₁N₃)₂·H₂SO₄·xH₂O
Storage Temperature: Room Temperature
MDL Number: MFCD00149650
CAS-Nummer: 1811-28-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=793e18138ad1dcd9bab9827c5b4ea0a650f11c67&vid=793e18138ad1dcd9bab9827c5b4ea0a650f11c67&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Faint Orange to Red-Brown to Brown
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Water (by Karl Fischer) ≤10.0 %
Solubility (Colour) Faint Orange to Dark Brown
Solubility (Form)
10 mg/ml, H2O
Solution
Carbon
Anhydrous
59.2 - 61.7 %
Nitrogen
Anhydrous
15.9 - 16.6 %

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge