Sigma Pseudo™ Corpse Scent, Formulation II, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P3929-20ML P3929-4X1ML P3929-8X1ML
P3929-20MLEA 255 EUR InStock
P3929-20ML P3929-4X1ML P3929-8X1ML
Sigma Pseudo™ Corpse Scent, Formulation II, Supelco®
Standards Organische Standards
Canine training aids for the detection of corpses. For post-putrification detection.
Sigma pseudo™ corpse scent may be used as an effective artificial scent in training cadaver dogs. The volatile compounds of its decomposition have been analyzed using gas chromatography coupled with time of flight mass spectrometry (GC–TOFMS).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge