Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ferrozine mono-sodium salt hydrate, BioXtra, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


M-Clarity™ Qualitätsstufe = MQ200

P5338-1G P5338-250MG P5338-5G
P5338-1GEA 213 EUR InStock
P5338-1G P5338-250MG P5338-5G
Ferrozine mono-sodium salt hydrate, BioXtra, Sigma-Aldrich®
Ferrozine mono-sodium salt hydrate
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4"-disulfonic acid sodium salt is also known as ferrozine.

  • 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4"-disulfonic acid sodium salt (ABTS) has been used in the ferene-s assay for the accurate determination of intracellular iron from iron oxide nanoparticles
  • It has also been used in ABTS radical scavenging assay to determine the antioxidant activity of honey samples


Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
Melting Pt: ≥300 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00078417
CAS-Nummer: 63451-29-6
EINECS: 274-196-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b37f37e9f5a144e9b4b635ee4e45bcf3d60db43&vid=6b37f37e9f5a144e9b4b635ee4e45bcf3d60db43&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Yellow to Yellow-Green
Appearance (Form) Powder
Insoluble matter ≤0.1 %
Chloride Content ≤0.1 %
Aluminum (AL) ≤0.001 %
Phosphorus (P) ≤0.0005 %
Lead (Pb) ≤0.001 %
Iron (Fe) ≤0.0005 %
Copper (Cu) ≤0.0005 %
Zinc (Zn) ≤0.0005 %
Calcium (Ca) ≤0.01 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.005 %

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge