Tablet count kits for water hardness
Lieferant: PALINTEST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AD-1016
PALIAD-1016EA 450 EUR InStock
PALIAD-1016
Tablet count kits for water hardness
Umwelt-Prüfkits
Tablet Count kits provide a simple method of analysis for an important range of water parameters. The tests are particularly useful for the routine control of industrial waters such as boiler waters and cooling water systems, as well as swimming pools, natural waters and similar applications.
  • Cost effective and provides quick results on-site
  • Can be used as a stand alone kit or combined with comparator or photometer
  • Wide range of test concentrations can be measured
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge