Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Depth filter capsules, Supracap™ 50

Lieferant: Pall Laboratory

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

SC050PDK5 SC050XAK7 SC050B020 SC050P900 SC050PDD1 SC050PDE2 SC050PDH4 SC050P100 SC050P200 SC050P250 SC050P700
PALLSC050PDK5EA 46.7 EUR InStock
PALLSC050PDK5 PALLSC050XAK7 PALLSC050B020 PALLSC050P900 PALLSC050PDD1 PALLSC050PDE2 PALLSC050PDH4 PALLSC050P100 PALLSC050P200 PALLSC050P250 PALLSC050P700
Depth filter capsules, Supracap™ 50
Filter Membranfilterkapseln
Scalable capsules with PP housing and vent and Seitz® high performance depth filter media. Supracap™ 50 capsules are designed for developing and optimising a process during scale-up and scale-down studies. They can be used to quickly and accurately determine which series and grade of depth filter media will provide the best performance as well as the necessary filtration area required to meet process volume.

  • Autoclavable at 125 °C
  • Low hold-up volume
  • Batch tested in order to meet all quality requirements

Supracap™ 50 capsules are readily scalable to Pall Supracap™ 100 capsules, Supradisc™ I, and Supradisc II depth filter modules.

Fields of application: Cell harvesting, clarification of fermentation broth, antibiotics, serum, blood products, vaccines, plant extracts.

The Seitz® Bio 20 media grade is made of highly purified natural and modified celluloses and is free from inorganic materials. Seitz® Bio 20 media is ideal for ion sensitive products such as therapeutic proteins, dialysis solutions, and protein-based diagnostics.

Seitz® P-series media are comprised of cellulose fibers, filter aids (diatomaceous earth and perlite), and resins. This grade is recommended for coarse filtration in the biotechnological and pharmaceutical industries.

Seitz® HP-series media are feature two distinct layers of Seitz® P-series depth filter sheets that feature a more permeable layer followed by a less permeable layer (PDD1, PDE2, PDH4 and PDK5). Seitz® HP-series media performs exceptionally well in low viability, high solids-containing applications, and with wider distribution of particle sizes in biotech applications.
The Seitz® AKS7 media grade is made of activated carbon within a matrix of cellulosic fibers. This immobilized carbon media is then coupled with a downstream filter paper to prevent carbon particle shedding downstream of the filter. The Seitz® AKS7 media grade is recommended for decolorisation and removal of other trace impurities.

Zertifizierungen: Manufactured under a Quality Management System certified to ISO 9000. All plastic components used in construction meet the specifications for Biological Reactivity Tests in vivo for Class VI Plastics (121 °C) as described in the current United States Pharmacopoeia (USP).

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge