Human plasma, gastric cancer (GC)
Lieferant: HEMACARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB006C-1-GC
PB006C-1-GCEA 0 EUR InStock
PB006C-1-GC
Human plasma, gastric cancer (GC)
Plasma
Plasma used for researching the causes, diagnosis, supportive care and treatment of gastric cancer (GC). Patient samples can be used by researchers to investigate biomarker and diagnostic development, pre-clinical research validation, and drug toxicity and efficacy. Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of gastric cancer (GC). Collected from IRB-consented donors diagnosed with GC.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Achtung! Bitte beachten: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge