Human mononuclear cells, DM2 donors
Lieferant: HEMACARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB009C-1-DM2
PB009C-1-DM2EA 0 EUR InStock
PB009C-1-DM2
Human mononuclear cells, DM2 donors
Zellen
Mononuclear cells used for researching the causes, diagnosis, supportive care and treatment of Diabetes Mellitus 2 (DM2), the most common form of diabetes. Human mononuclear cells are collected from human volunteer donors diagnosed with DM2 under IRB approved informed consent by whole blood or leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol.
  • Cryopreserved in CryoStor® CS10 media
Donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80 °C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.
Achtung! Bitte beachten: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge