Human mononuclear cells from peripheral blood, Hepatitis C virus (HCV)
Lieferant: HEMACARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PB009C-1-HCV
PB009C-1-HCVEA 0 EUR InStock
PB009C-1-HCV
Human mononuclear cells from peripheral blood, Hepatitis C virus (HCV)
Zellen
Mononuclear cells (MNCs) used for researching the causes, diagnosis, supportive care and treatment of Hepatitis C Virus (HCV). Human mononuclear cells are collected from human volunteer donors diagnosed with Hepatitis C virus under IRB approved informed consent.
  • Cryopreserved in CryoStor® CS10 media
Storage: Either prepare cells for long term storage in liquid nitrogen vapour phase or thaw for use. Storage in liquid phase nitrogen is NOT recommended. Short term storage of cells (less than 2 weeks) at –80 °C is acceptable, but should be minimised to ensure maximum stability. Once thawed, samples must be used immediately.
Achtung! Bitte beachten: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge