IQ/OQ certificaton for gas generators
Lieferant: PEAK SCIENTIFIC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
09-2100
PEAK09-2100EA 0 EUR InStock
PEAK09-2100
IQ/OQ certificaton for gas generators
Services
With customer service and support in mind a comprehensive Installation and Operational Qualification certification has been developed, for customers who require this additional certified assurance for their analytical applications.
  • IQ/OQ is tailored to the specific generator
  • Generator compliance certification for regulated applications
  • Carried out at initial installation or equipment relocation
IQ is the pre-installation detail of the equipment, its component parts and its location onsite.
OQ is to verify that the equipment functions according to the generator’s specifications.
This provides a cost-effective and efficient solution to achieve and maintain compliance. In addition to our highly trained Service Engineer completing the Installation and Operational validation, we will also provide full certification to document this process, ensuring accurate proof of compliance.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge